4-(tert-Butyl)-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

53773-500MG 53773-1G 53773-5G 53773-25G
77329-110EA 82.4 USD
77329-110 77342-152 77365-406 77377-134
4-(tert-Butyl)-2-methoxybenzoic acid ≥97%
4-(tert-Butyl)-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD07778625
CAS Number: 52328-48-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR