1-Bromo-4-(tert-butyl)-2-(difluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

53767-500MG 53767-1G 53767-5G 53767-25G
77329-108EA 234.84 USD
77329-108 77352-912 77365-404 77387-416
1-Bromo-4-(tert-butyl)-2-(difluoromethoxy)benzene ≥97%
1-Bromo-4-(tert-butyl)-2-(difluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrF₂O
MW: 279.121 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757133
CAS Number: 2764728-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR