(4,5-Difluoro-2-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

53760-500MG 53760-1G 53760-5G 53760-25G
77329-106EA 266.77 USD
77329-106 77352-910 77365-402 77387-414
(4,5-Difluoro-2-methoxyphenyl)(methyl)sulfane ≥97%
(4,5-Difluoro-2-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈F₂OS
Storage Temperature: Ambient
MDL Number: MFCD26960277
CAS Number: 1660115-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR