2-Ethoxy-4,5-difluorophenol ≥97%

Supplier: AOBChem USA

Warning

53759-500MG 53759-1G 53759-5G 53759-25G
77329-104EA 201.88 USD
77329-104 77352-908 77365-400 77387-412
2-Ethoxy-4,5-difluorophenol ≥97%
2-Ethoxy-4,5-difluorophenol

Precaución: for Research Use Only

Formula: C₈H₈F₂O₂
MW: 174.145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757128
CAS Number: 2091449-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR