1-Ethoxy-4,5-difluoro-2-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

53758-500MG 53758-1G 53758-5G 53758-25G
77329-102EA 168.92 USD
77329-102 77346-272 77365-398 77387-410
1-Ethoxy-4,5-difluoro-2-iodobenzene ≥97%
1-Ethoxy-4,5-difluoro-2-iodobenzene

Precaución: for Research Use Only

Formula: C₈H₇F₂IO
Storage Temperature: Ambient
MDL Number: MFCD34757127
CAS Number: 2090513-06-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR