(4,5-Difluoro-2-isopropoxyphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

53756-500MG 53756-1G 53756-5G 53756-25G
77329-098EA 189.52 USD
77329-098 77352-904 77365-396 77387-406
(4,5-Difluoro-2-isopropoxyphenyl)(methyl)sulfane ≥95%
(4,5-Difluoro-2-isopropoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₂F₂OS
MW: 218.263 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757126
CAS Number: 2757730-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR