1-(2,3,4-Trifluorophenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

53742-500MG 53742-1G 53742-5G 53742-25G
77329-096EA 242.05 USD
77329-096 77352-902 77365-394 77387-404
1-(2,3,4-Trifluorophenyl)ethanol ≥97%
1-(2,3,4-Trifluorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₇F₃O
MW: 176.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17254414
CAS Number: 1340441-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR