3-Fluoro-2-methyl-6-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

53738-500MG 53738-1G 53738-5G
77338-002EA 457.32 USD
77338-002 77352-898 77377-128
3-Fluoro-2-methyl-6-(methylthio)benzoic acid ≥97%
3-Fluoro-2-methyl-6-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₂S
Storage Temperature: Ambient
MDL Number: MFCD34757120
CAS Number: 2807445-23-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR