Methyl(4-propoxyphenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

53695-500MG 53695-1G 53695-5G 53695-25G
77329-090EA 99.91 USD
77329-090 77342-824 77365-386 77377-118
Methyl(4-propoxyphenyl)sulfane ≥95%
Methyl(4-propoxyphenyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₄OS
MW: 182.283 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564242
CAS Number: 178987-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR