1-(3,5-Dichlorophenyl)-2,2,2-trifluoroethan-1-ol ≥95%
Supplier: AOBChem USA
|
Warning
|
53679-500MG
53679-1G
53679-5G
53679-25G
77329-088EA
213.21
USD
77329-088
77352-888
77365-384
77387-398
1-(3,5-Dichlorophenyl)-2,2,2-trifluoroethan-1-ol ≥95%
1-(3,5-Dichlorophenyl)-2,2,2-trifluoroethan-1-ol
Precaución: for Research Use Only
Formula:
C₈H₅Cl₂F₃O Storage Temperature: Ambient |
MDL Number:
MFCD16067948 CAS Number: 1187989-89-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...