2-Fluoro-4-methoxy-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

53674-500MG 53674-1G 53674-5G 53674-25G
77329-086EA 196.73 USD
77329-086 77352-884 77365-382 77387-396
2-Fluoro-4-methoxy-3-methylbenzoic acid ≥97%
2-Fluoro-4-methoxy-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₃
MW: 184.164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22414700
CAS Number: 1427433-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR