5-(Benzyloxy)-2-chlorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

53661-500MG 53661-1G 53661-5G 53661-25G
77329-080EA 270.89 USD
77329-080 77352-874 77365-376 77387-390
5-(Benzyloxy)-2-chlorobenzaldehyde ≥97%
5-(Benzyloxy)-2-chlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₁ClO₂
Storage Temperature: Ambient
MDL Number: MFCD19303309
CAS Number: 876300-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR