5-(Benzyloxy)-2-chlorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

53660-500MG 53660-1G 53660-5G
77329-078EA 307.97 USD
77329-078 77352-872 77377-108
5-(Benzyloxy)-2-chlorobenzoic acid ≥97%
5-(Benzyloxy)-2-chlorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₁ClO₃
MW: 262.688 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12913366
CAS Number: 39062-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR