2-(3,4,5-Trifluorophenyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

53653-500MG 53653-1G 53653-5G 53653-25G
77329-074EA 206 USD
77329-074 77352-868 77365-372 77387-386
2-(3,4,5-Trifluorophenyl)propan-2-ol ≥97%
2-(3,4,5-Trifluorophenyl)propan-2-ol

Precaución: for Research Use Only

Formula: C₉H₉F₃O
MW: 190.162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20528502
CAS Number: 1379364-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR