1-Bromo-4-((2-fluorobenzyl)oxy)-2-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

53648-1G 53648-5G 53648-25G
77341-024EA 66.95 USD
77341-024 77365-370 77377-106
1-Bromo-4-((2-fluorobenzyl)oxy)-2-methoxybenzene ≥97%
1-Bromo-4-((2-fluorobenzyl)oxy)-2-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFO₂
MW: 311.146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757106
CAS Number: 2764730-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR