1-((4-Bromobenzyl)oxy)-3-fluoro-5-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

53646-1G 53646-5G 53646-25G
77341-284EA 81.37 USD
77341-284 77365-368 77377-104
1-((4-Bromobenzyl)oxy)-3-fluoro-5-(trifluoromethyl)benzene ≥97%
1-((4-Bromobenzyl)oxy)-3-fluoro-5-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₄H₉BrF₄O
MW: 349.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530225
CAS Number: 2734779-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR