2-Fluoro-3-methoxy-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

53619-500MG 53619-1G 53619-5G 53619-25G
77329-070EA 234.84 USD
77329-070 77352-864 77365-364 77387-384
2-Fluoro-3-methoxy-5-methylbenzoic acid ≥97%
2-Fluoro-3-methoxy-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₃
MW: 184.164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11110176
CAS Number: 447463-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR