(2-Fluoro-3-methoxy-5-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

53616-500MG 53616-1G 53616-5G 53616-25G
77329-068EA 230.72 USD
77329-068 77352-862 77365-362 77387-382
(2-Fluoro-3-methoxy-5-methylphenyl)(methyl)sulfane ≥97%
(2-Fluoro-3-methoxy-5-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁FOS
MW: 186.246 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28130824
CAS Number: 1452478-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR