2-Fluoro-1-methoxy-3,5-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

53614-500MG 53614-1G 53614-5G
77337-718EA 438.78 USD
77337-718 77352-860 77377-100
2-Fluoro-1-methoxy-3,5-dimethylbenzene ≥97%
2-Fluoro-1-methoxy-3,5-dimethylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD28794227
CAS Number: 1253214-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR