1-Bromo-5-ethyl-2-fluoro-3-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

53611-500MG 53611-1G 53611-5G 53611-25G
77329-064EA 234.84 USD
77329-064 77352-858 77365-358 77387-378
1-Bromo-5-ethyl-2-fluoro-3-iodobenzene ≥95%
1-Bromo-5-ethyl-2-fluoro-3-iodobenzene

Precaución: for Research Use Only

Formula: C₈H₇BrFI
Storage Temperature: Ambient
MDL Number: MFCD34757100
CAS Number: 2705600-39-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR