1-Bromo-5-ethyl-2-fluoro-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

53610-500MG 53610-1G 53610-5G
77336-628EA 390.37 USD
77336-628 77352-856 77377-098
1-Bromo-5-ethyl-2-fluoro-3-methylbenzene ≥97%
1-Bromo-5-ethyl-2-fluoro-3-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrF
Storage Temperature: Ambient
MDL Number: MFCD34757099
CAS Number: 2710513-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR