(3-Iodo-5-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

53604-500MG 53604-1G 53604-5G 53604-25G
77329-062EA 184.37 USD
77329-062 77347-158 77365-356 77387-376
(3-Iodo-5-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%
(3-Iodo-5-(trifluoromethyl)phenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₆F₃IS
MW: 318.098 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22488938
CAS Number: 1822630-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR