2-Chloro-6-isopropylphenol ≥97%

Supplier: AOBChem USA

Warning

53582-500MG 53582-1G 53582-5G
77329-060EA 307.97 USD
77329-060 77352-854 77377-096
2-Chloro-6-isopropylphenol ≥97%
2-Chloro-6-isopropylphenol

Precaución: for Research Use Only

Formula: C₉H₁₁ClO
Storage Temperature: Ambient
MDL Number: MFCD16997222
CAS Number: 57883-02-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR