1-Bromo-3-isopropyl-2-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

53579-500MG 53579-1G 53579-5G 53579-25G
77329-058EA 235.87 USD
77329-058 77352-852 77365-354 77387-374
1-Bromo-3-isopropyl-2-methoxybenzene ≥97%
1-Bromo-3-isopropyl-2-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrO
MW: 229.114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11845672
CAS Number: 129976-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR