1-Bromo-2-ethoxy-3-isopropylbenzene ≥97%

Supplier: AOBChem USA

Warning

53578-500MG 53578-1G 53578-5G 53578-25G
77329-056EA 236.9 USD
77329-056 77352-850 77365-352 77387-372
1-Bromo-2-ethoxy-3-isopropylbenzene ≥97%
1-Bromo-2-ethoxy-3-isopropylbenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrO
MW: 243.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757079
CAS Number: 2755717-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR