5-Fluoro-N,2-dimethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

53574-1G 53574-5G 53574-25G
77341-022EA 66.95 USD
77341-022 77365-350 77377-094
5-Fluoro-N,2-dimethylbenzamide ≥97%
5-Fluoro-N,2-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₉H₁₀FNO
MW: 167.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19596596
CAS Number: 1332325-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR