N-Cyclopropyl-5-fluoro-2-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

53573-1G 53573-5G 53573-25G
77341-154EA 72.1 USD
77341-154 77365-348 77377-092
N-Cyclopropyl-5-fluoro-2-methylbenzamide ≥97%
N-Cyclopropyl-5-fluoro-2-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₂FNO
Storage Temperature: Ambient
MDL Number: MFCD20356322
CAS Number: 1344222-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR