(5-Fluoro-2-methylphenyl)(4-methylpiperazin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

53569-1G 53569-5G 53569-25G
77341-928EA 108.15 USD
77341-928 77365-340 77377-084
(5-Fluoro-2-methylphenyl)(4-methylpiperazin-1-yl)methanone ≥97%
(5-Fluoro-2-methylphenyl)(4-methylpiperazin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₇FN₂O
MW: 236.285 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20044469
CAS Number: 864054-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR