1-Bromo-2-iodo-5-methoxy-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

53564-500MG 53564-1G 53564-5G 53564-25G
77329-050EA 141.11 USD
77329-050 77344-832 77365-336 77387-370
1-Bromo-2-iodo-5-methoxy-4-methylbenzene ≥97%
1-Bromo-2-iodo-5-methoxy-4-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈BrIO
MW: 326.957 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31672671
CAS Number: 2386922-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR