4-Bromo-2-fluoro-5-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

53563-500MG 53563-1G 53563-5G 53563-25G
77329-048EA 247.2 USD
77329-048 77352-848 77365-334 77387-368
4-Bromo-2-fluoro-5-hydroxybenzaldehyde ≥97%
4-Bromo-2-fluoro-5-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD17012742
CAS Number: 943750-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR