4-Fluoro-2-hydroxy-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

53553-500MG 53553-1G 53553-5G
77329-046EA 322.39 USD
77329-046 77352-846 77377-080
4-Fluoro-2-hydroxy-5-methylbenzaldehyde ≥97%
4-Fluoro-2-hydroxy-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇FO₂
MW: 154.138 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18824465
CAS Number: 504414-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR