Ethyl 2-bromo-4-fluoro-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

53547-500MG 53547-1G 53547-5G 53547-25G
77329-044EA 141.11 USD
77329-044 77344-830 77365-332 77387-366
Ethyl 2-bromo-4-fluoro-5-methylbenzoate ≥97%
Ethyl 2-bromo-4-fluoro-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD28739703
CAS Number: 1807168-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR