3-Bromo-6-chloro-2-methyl-5-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

53538-1G 53538-5G 53538-25G
77342-018EA 111.24 USD
77342-018 77365-326 77377-078
3-Bromo-6-chloro-2-methyl-5-(methylthio)pyridine ≥97%
3-Bromo-6-chloro-2-methyl-5-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₇H₇BrClNS
MW: 252.559 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757063
CAS Number: 2764730-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR