3,5-Dibromo-2-methoxy-6-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

53530-500MG 53530-1G 53530-5G 53530-25G
77329-036EA 147.29 USD
77329-036 77345-208 77365-322 77387-358
3,5-Dibromo-2-methoxy-6-methylpyridine ≥97%
3,5-Dibromo-2-methoxy-6-methylpyridine

Precaución: for Research Use Only

Formula: C₇H₇Br₂NO
MW: 280.945 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185523
CAS Number: 156094-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR