3-Bromo-6-fluoro-5-iodo-2-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

53522-1G 53522-5G 53522-25G
77341-826EA 105.06 USD
77341-826 77365-320 77377-076
3-Bromo-6-fluoro-5-iodo-2-methylpyridine ≥97%
3-Bromo-6-fluoro-5-iodo-2-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₄BrFIN
Storage Temperature: Ambient
MDL Number: MFCD34757054
CAS Number: 2764733-61-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR