Methyl 4-ethoxy-2-fluoro-5-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

53516-500MG 53516-1G 53516-5G
77337-282EA 410.97 USD
77337-282 77352-840 77377-072
Methyl 4-ethoxy-2-fluoro-5-methoxybenzoate ≥97%
Methyl 4-ethoxy-2-fluoro-5-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₄
MW: 228.217 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28360382
CAS Number: 2383800-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR