4-Ethoxy-2-fluoro-5-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

53515-500MG 53515-1G 53515-5G
77336-626EA 390.37 USD
77336-626 77352-838 77377-070
4-Ethoxy-2-fluoro-5-methoxybenzoic acid ≥97%
4-Ethoxy-2-fluoro-5-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₄
Storage Temperature: Ambient
MDL Number: MFCD28360381
CAS Number: 2167188-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR