1-Ethoxy-5-fluoro-4-iodo-2-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

53513-500MG 53513-1G 53513-5G
77329-034EA 350.2 USD
77329-034 77352-834 77377-066
1-Ethoxy-5-fluoro-4-iodo-2-methoxybenzene ≥97%
1-Ethoxy-5-fluoro-4-iodo-2-methoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀FIO₂
Storage Temperature: Ambient
MDL Number: MFCD34564239
CAS Number: 2092536-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR