5-(3-Formylphenyl)-N-methylpicolinamide ≥97%

Supplier: AOBChem USA

Warning

53505-500MG 53505-1G 53505-5G
77339-018EA 544.87 USD
77339-018 77352-832 77377-064
5-(3-Formylphenyl)-N-methylpicolinamide ≥97%
5-(3-Formylphenyl)-N-methylpicolinamide

Precaución: for Research Use Only

Formula: C₁₄H₁₂N₂O₂
MW: 240.257 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178707
CAS Number: 2643368-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR