3-(5-Ethoxypyridin-3-yl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

53504-500MG 53504-1G 53504-5G 53504-25G
77329-032EA 202.91 USD
77329-032 77352-830 77365-318 77387-356
3-(5-Ethoxypyridin-3-yl)benzaldehyde ≥97%
3-(5-Ethoxypyridin-3-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₃NO₂
MW: 227.259 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25743914
CAS Number: 1546153-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR