5-(3-Formylphenyl)-N,N-dimethylpicolinamide ≥97%

Supplier: AOBChem USA

Warning

53503-500MG 53503-1G 53503-5G
77338-650EA 506.76 USD
77338-650 77352-828 77377-062
5-(3-Formylphenyl)-N,N-dimethylpicolinamide ≥97%
5-(3-Formylphenyl)-N,N-dimethylpicolinamide

Precaución: for Research Use Only

Formula: C₁₅H₁₄N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34178706
CAS Number: 2643368-37-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR