3-(6-Fluoro-5-methylpyridin-3-yl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

53499-500MG 53499-1G 53499-5G
77337-884EA 450.11 USD
77337-884 77352-826 77377-060
3-(6-Fluoro-5-methylpyridin-3-yl)benzaldehyde ≥95%
3-(6-Fluoro-5-methylpyridin-3-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₀FNO
MW: 215.223 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178705
CAS Number: 2643367-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR