2-(Benzyloxy)-3-chloro-4-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

53491-500MG 53491-1G 53491-5G
77337-428EA 414.06 USD
77337-428 77352-824 77377-058
2-(Benzyloxy)-3-chloro-4-(methylthio)pyridine ≥97%
2-(Benzyloxy)-3-chloro-4-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₁₃H₁₂ClNOS
MW: 265.758 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757048
CAS Number: 2764728-07-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR