2-(Benzyloxy)-3-chloro-4-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

53489-500MG 53489-1G 53489-25G 53489-5G
77329-030EA 297.67 USD
77329-030 77352-822 77387-354 77377-056
2-(Benzyloxy)-3-chloro-4-iodopyridine ≥97%
2-(Benzyloxy)-3-chloro-4-iodopyridine

Precaución: for Research Use Only

Formula: C₁₂H₉ClINO
Storage Temperature: Ambient
MDL Number: MFCD34564238
CAS Number: 2734776-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR