3-Chloro-2-methoxy-4-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

53488-1G 53488-5G 53488-25G
77341-282EA 81.37 USD
77341-282 77365-316 77377-054
3-Chloro-2-methoxy-4-(methylthio)pyridine ≥97%
3-Chloro-2-methoxy-4-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₇H₈ClNOS
MW: 189.663 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757047
CAS Number: 2764728-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR