3-Bromo-4-fluorobenzyl fluoride ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-1-fluoro-4-(fluoromethyl)benzene

53464-500MG 53464-1G 53464-5G
77337-034EA 406.85 USD
77337-034 77352-812 77377-046
3-Bromo-4-fluorobenzyl fluoride ≥95%
3-Bromo-4-fluorobenzyl fluoride

Precaución: for Research Use Only

Formula: C₇H₅BrF₂
MW: 207.015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28527820
CAS Number: 1783551-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR