(4-Methoxy-2,6-dimethylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

53454-1G 53454-5G 53454-25G
77341-428EA 85.49 USD
77341-428 77365-312 77377-040
(4-Methoxy-2,6-dimethylphenyl)(methyl)sulfane ≥97%
(4-Methoxy-2,6-dimethylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₄OS
MW: 182.283 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28795063
CAS Number: 55661-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR