(3-Bromo-2-fluoro-5,6-dimethoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

53451-500MG 53451-1G 53451-5G 53451-25G
77329-022EA 278.1 USD
77329-022 77352-806 77365-308 77387-348
(3-Bromo-2-fluoro-5,6-dimethoxyphenyl)methanol ≥95%
(3-Bromo-2-fluoro-5,6-dimethoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₀BrFO₃
MW: 265.076 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178702
CAS Number: 1898413-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR