3-Bromo-2-fluoro-5,6-dimethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

53450-500MG 53450-1G 53450-5G
77329-020EA 310.03 USD
77329-020 77352-804 77377-036
3-Bromo-2-fluoro-5,6-dimethoxybenzaldehyde ≥97%
3-Bromo-2-fluoro-5,6-dimethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈BrFO₃
MW: 263.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178701
CAS Number: 1785348-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR