1-Bromo-2-fluoro-4,5-dimethoxy-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

53447-500MG 53447-1G 53447-5G 53447-25G
77329-018EA 166.86 USD
77329-018 77346-212 77365-306 77387-346
1-Bromo-2-fluoro-4,5-dimethoxy-3-methylbenzene ≥97%
1-Bromo-2-fluoro-4,5-dimethoxy-3-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrFO₂
MW: 249.077 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178699
CAS Number: 2643367-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR